ENA238711054 Enamine

1-cyclopentyl-1-phenylmethanamine hydrochloride

  • MDL number MFCD12197063
  • Cas Number : 24260-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44055
Availability details for
Quantity
  • Availability
  • Add to Cart