ENA238711151 Enamine

4-(chloromethyl)-2-nitrothiophene

  • MDL number MFCD12197117
  • Cas Number : 1092561-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-44872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44872
Availability details for
Quantity
  • Availability
  • Add to Cart