ENA238711156 Enamine

tert-butyl N-{1-[(1H-indol-3-yl)methyl]piperidin-4-yl}carbamate

  • MDL number MFCD12197133
  • Cas Number : 1181458-58-1
  • Molecular Weight : 329.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44958
Availability details for
Quantity
  • Availability
  • Add to Cart