ENA238711157 Enamine

2-(4-chlorophenyl)-2-(methylamino)acetamide

  • MDL number MFCD12197136
  • Cas Number : 71972-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-154
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44990

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-44990
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart