ENA238711174 Enamine

3-fluoro-4-(1H-pyrazol-1-yl)aniline dihydrochloride

  • MDL number MFCD12197213
  • Cas Number : 1193387-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45469
Availability details for
Quantity
  • Availability
  • Add to Cart