ENA238711176 Enamine

1-(2-bromoethyl)-2-methyl-1H-1,3-benzodiazole hydrobromide

  • MDL number MFCD12197218
  • Cas Number : 1193390-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-45482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45482
Availability details for
Quantity
  • Availability
  • Add to Cart