ENA238711177 Enamine

4-chloro-1-ethyl-1H-imidazole-5-carbaldehyde

  • MDL number MFCD12197219
  • Cas Number : 1193388-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45499

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45499
Availability details for
Quantity
  • Availability
  • Add to Cart