ENA238711178 Enamine

ethyl 5-(chlorosulfonyl)-3-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD12197220
  • Cas Number : 1299471-28-5
  • Molecular Weight : 252.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-(chlorosulfonyl)-3-methyl-1H-pyrazole-4-carboxylate
Inchi Code 1S/C7H9ClN2O4S/c1-3-14-7(11)5-4(2)9-10-6(5)15(8,12)13/h3H2,1-2H3,(H,9,10)
Inchi Key YBVUJZKYBUCSOY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-45515

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-45515
pictograms GHS05,GHS07
hstatements H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart