ENA238711182 Enamine

2-amino-6-chlorobenzene-1-thiol hydrochloride

  • MDL number MFCD12197228
  • Cas Number : 385376-58-9
  • Molecular Weight : 196.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-45569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45569
Availability details for
Quantity
  • Availability
  • Add to Cart