ENA238711187 Enamine

4,5-dimethyl 1-(piperidin-4-yl)-1H-1,2,3-triazole-4,5-dicarboxylate hydrochloride

  • MDL number MFCD12197254
  • Cas Number : 1193389-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49609
Availability details for
Quantity
  • Availability
  • Add to Cart