ENA238711188 Enamine

4-amino-N-{4-[ethyl(2-hydroxyethyl)amino]phenyl}benzene-1-sulfonamide

  • MDL number MFCD12912776
  • Cas Number : 1193389-97-7
  • Molecular Weight : 335.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49629
Availability details for
Quantity
  • Availability
  • Add to Cart