ENA238711189 Enamine

methyl 5-(4-aminobenzenesulfonamido)-2-chlorobenzoate

  • MDL number MFCD12912777
  • Cas Number : 1193390-00-9
  • Molecular Weight : 340.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49632
Availability details for
Quantity
  • Availability
  • Add to Cart