ENA238711191 Enamine

3-(2,2,2-trifluoro-1,1-dihydroxyethyl)-4H-chromen-4-one

  • MDL number MFCD12197262
  • Cas Number : 858369-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49729
Availability details for
Quantity
  • Availability
  • Add to Cart