ENA238711196 Enamine

ethyl (2Z)-2-cyano-3-[(2-fluorophenyl)amino]prop-2-enoate

  • MDL number MFCD00855212
  • Cas Number : 934070-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49740

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49740
Availability details for
Quantity
  • Availability
  • Add to Cart