ENA238711198 Enamine

4-(2,2,2-trifluoroethoxy)butan-1-amine hydrochloride

  • MDL number MFCD12197268
  • Cas Number : 1193389-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49744
Availability details for
Quantity
  • Availability
  • Add to Cart