ENA238711200 Enamine

ethyl (2Z)-2-cyano-3-[(2,5-difluorophenyl)amino]prop-2-enoate

  • MDL number MFCD00113374
  • Cas Number : 239085-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49748

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49748
Availability details for
Quantity
  • Availability
  • Add to Cart