ENA238711217 Enamine

3-amino-5-fluorobenzene-1-sulfonamide

  • MDL number MFCD12197387
  • Cas Number : 1193388-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50125
Availability details for
Quantity
  • Availability
  • Add to Cart