ENA238711222 Enamine

1-(4-chlorophenyl)-4-methylpentan-1-amine

  • MDL number MFCD11116711
  • Cas Number : 161531-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44253
Availability details for
Quantity
  • Availability
  • Add to Cart