ENA238711230 Enamine

7-(hydroxymethyl)-3-methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one

  • MDL number MFCD08442964
  • Cas Number : 926257-63-8
  • Molecular Weight : 196.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44273
Availability details for
Quantity
  • Availability
  • Add to Cart