ENA238711232 Enamine

5-(ethylsulfamoyl)-2-fluorobenzoic acid

  • MDL number MFCD03765569
  • Cas Number : 632300-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44278

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44278
Availability details for
Quantity
  • Availability
  • Add to Cart