ENA238711234 Enamine

8a-methyl-5-oxo-hexahydro-[1,3]thiazolo[2,3-c]thiomorpholine-3-carboxylic acid

  • MDL number MFCD11858190
  • Cas Number : 1214654-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44281
Availability details for
Quantity
  • Availability
  • Add to Cart