ENA238711237 Enamine

5,6-dichloropyridine-3-carboxamide

  • MDL number MFCD00052827
  • Cas Number : 75291-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44287

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44287
Availability details for
Quantity
  • Availability
  • Add to Cart