ENA238711243 Enamine

3-[3-(thiophen-2-yl)prop-2-enamido]benzoic acid

  • Cas Number : 1016753-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44300
Availability details for
Quantity
  • Availability
  • Add to Cart