ENA238711245 Enamine

4-methylquinoline-2-carboxylic acid

  • MDL number MFCD02230093
  • Cas Number : 40609-76-5
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-44310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44310
Availability details for
Quantity
  • Availability
  • Add to Cart