ENA238711248 Enamine

2-(prop-2-en-1-yloxy)acetic acid

  • MDL number MFCD10696056
  • Cas Number : 22577-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-44821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44821
Availability details for
Quantity
  • Availability
  • Add to Cart