ENA238711259 Enamine

{3-[(oxolan-2-yl)methoxy]phenyl}methanamine

  • MDL number MFCD08690186
  • Cas Number : 919017-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-44842

Safety Information

signalword Danger
pcodes P280,P305+P351+P338,P310
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-44842
pictograms GHS05
hstatements H318
Availability details for
Quantity
  • Availability
  • Add to Cart