ENA238711262 Enamine

4-bromo-2-(pyrrolidine-1-carbonyl)-1H-pyrrole

  • MDL number MFCD10635100
  • Cas Number : 1031501-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44847
Availability details for
Quantity
  • Availability
  • Add to Cart