ENA238711273 Enamine

4-{[(2-methylphenyl)methyl]amino}butanoic acid hydrochloride

  • MDL number MFCD12197116
  • Cas Number : 1181458-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44868
Availability details for
Quantity
  • Availability
  • Add to Cart