ENA238711274 Enamine

1-(4-phenylphenyl)propan-2-amine hydrochloride

  • MDL number MFCD01740505
  • Cas Number : 82804-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-238
Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44870
Availability details for
Quantity
  • Availability
  • Add to Cart