ENA238711275 Enamine

5-[(4-chloro-2-methylphenyl)methyl]-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD12197069
  • Cas Number : 1171404-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44080
Availability details for
Quantity
  • Availability
  • Add to Cart