ENA238711282 Enamine

2-(pyridine-3-amido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid

  • MDL number MFCD00705119
  • Cas Number : 314032-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44897
Availability details for
Quantity
  • Availability
  • Add to Cart