ENA238711283 Enamine

2-[(1,3-benzothiazol-2-yl)amino]acetic acid

  • MDL number MFCD00463455
  • Cas Number : 91192-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-44900

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P273,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-44900
pictograms GHS07
hstatements H302,H315,H319,H335,H412
Availability details for
Quantity
  • Availability
  • Add to Cart