ENA238711284 Enamine

4-(cyclopentyloxy)-3-methoxybenzoic acid

  • MDL number MFCD06199287
  • Cas Number : 176033-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44902

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44902
Availability details for
Quantity
  • Availability
  • Add to Cart