ENA238711286 Enamine

3-(4-ethylpiperazin-1-yl)-2-methylpropan-1-amine

  • MDL number MFCD09901921
  • Cas Number : 1017465-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44907
Availability details for
Quantity
  • Availability
  • Add to Cart