ENA238711304 Enamine

3-methyl-5-(2-phenoxyacetamido)thiophene-2-carboxylic acid

  • MDL number MFCD09812450
  • Cas Number : 1016535-37-7
  • Molecular Weight : 291.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methyl-5-[(phenoxyacetyl)amino]-2-thiophenecarboxylic acid
Inchi Code 1S/C14H13NO4S/c1-9-7-12(20-13(9)14(17)18)15-11(16)8-19-10-5-3-2-4-6-10/h2-7H,8H2,1H3,(H,15,16)(H,17,18)
Inchi Key AQFQTLPZZODVQE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-44947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44947
Availability details for
Quantity
  • Availability
  • Add to Cart