ENA238711312 Enamine

4-[4-(2-methylpropyl)phenyl]-1,3-thiazol-2-amine

  • MDL number MFCD01471574
  • Cas Number : 351982-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44961
Availability details for
Quantity
  • Availability
  • Add to Cart