ENA238711316 Enamine

5,6-dichloro-N-cyclopentylpyridine-3-carboxamide

  • MDL number MFCD09804622
  • Cas Number : 1016744-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44971
Availability details for
Quantity
  • Availability
  • Add to Cart