ENA238711325 Enamine

2-(1H-imidazol-1-yl)-1-[3-(trifluoromethyl)phenyl]ethan-1-amine dihydrochloride

  • MDL number MFCD12197138
  • Cas Number : 1181458-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 273...275
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44998

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44998
Availability details for
Quantity
  • Availability
  • Add to Cart