ENA238711328 Enamine

5-amino-1-propyl-1,2-dihydropyridin-2-one hydrochloride

  • MDL number MFCD12197140
  • Cas Number : 1181458-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-45006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45006
Availability details for
Quantity
  • Availability
  • Add to Cart