ENA238711349 Enamine

N1,4-dimethylbenzene-1,2-diamine

  • MDL number MFCD07437799
  • Cas Number : 39513-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45050
Availability details for
Quantity
  • Availability
  • Add to Cart