ENA238711355 Enamine

3-(2-methylphenyl)-1,2-oxazol-5-amine

  • MDL number MFCD07383027
  • Cas Number : 86685-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-45093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45093
Availability details for
Quantity
  • Availability
  • Add to Cart