ENA238711357 Enamine

methyl 2-methanesulfonamido-3-methylbutanoate

  • MDL number MFCD12197358
  • Cas Number : 322394-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45097
Availability details for
Quantity
  • Availability
  • Add to Cart