ENA238711367 Enamine

tert-butyl N-methyl-N-(piperidin-3-ylmethyl)carbamate

  • MDL number MFCD06800386
  • Cas Number : 169750-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45119

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45119
Availability details for
Quantity
  • Availability
  • Add to Cart