ENA238711369 Enamine

4-sulfamoyl-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid

  • MDL number MFCD09047325
  • Cas Number : 926233-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45127
Availability details for
Quantity
  • Availability
  • Add to Cart