ENA238711372 Enamine

4-[(4-methyl-1,3-thiazol-2-yl)sulfanyl]aniline

  • MDL number MFCD11132439
  • Cas Number : 1019365-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45135
Availability details for
Quantity
  • Availability
  • Add to Cart