ENA238711374 Enamine

3-[(2-methylpropyl)amino]propanoic acid hydrochloride

  • MDL number MFCD12197154
  • Cas Number : 1181458-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45139

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45139
Availability details for
Quantity
  • Availability
  • Add to Cart