ENA238711387 Enamine

3-(1H-1,2,4-triazol-1-ylmethyl)aniline

  • MDL number MFCD08060528
  • Cas Number : 127988-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45168
Availability details for
Quantity
  • Availability
  • Add to Cart