ENA238711395 Enamine

4-(2-oxo-1,3-oxazolidin-3-yl)butanoic acid

  • MDL number MFCD11131208
  • Cas Number : 933686-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-45182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45182
Availability details for
Quantity
  • Availability
  • Add to Cart