ENA238711396 Enamine

4-{[(3-fluorophenyl)methyl]amino}butanoic acid hydrochloride

  • MDL number MFCD12197162
  • Cas Number : 1181458-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45184
Availability details for
Quantity
  • Availability
  • Add to Cart