ENA238711399 Enamine

1-[4-(aminomethyl)-2-fluorophenyl]piperidin-4-ol

  • MDL number MFCD11191685
  • Cas Number : 1039892-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45190

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45190
Availability details for
Quantity
  • Availability
  • Add to Cart